Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026930.4 | NP_001022101.1 | 3 - 4 | 924 | 106 | |
NM_001026931.6 | NP_001022102.1 | 4 - 5 | 412 | 106 | ||
GenBank mRNA | AF083234.1 | - | 2 - 3 | 301 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |