Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063148.6 | NP_495549.1 | 4 - 5 | 420 | 132 | |
NM_063149.5 | NP_495550.1 | 4 - 5 | 399 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |