Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027334.3 | NP_001022505.1 | 2 - 3 | 260 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |