Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063258.8 | NP_495659.1 | 3 - 4 | 370 | 85 | |
GenBank mRNA | AB000919.1 | - | 3 - 4 | 381 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |