Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063296.6 | NP_495697.2 | 1 - 2 | 165 | 104 | |
GenBank mRNA | AF489839.1 | - | 1 - 2 | 187 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |