Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322746.3 | NP_001309573.1 | 1 - 2 | 133 | 131 | |
NM_063296.6 | NP_495697.2 | 4 - 5 | 900 | 131 | ||
GenBank mRNA | AF489839.1 | - | 4 - 5 | 922 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |