Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063315.3 | NP_495716.1 | 10 - 11 | 1265 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |