Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027088.7 | NP_001022259.1 | 4 - 5 | 403 | 112 | |
NM_001027089.7 | NP_001022260.1 | 3 - 4 | 274 | 112 | ||
NM_001373755.2 | NP_001359723.1 | 3 - 4 | 293 | 112 | ||
NM_001381521.1 | NP_001368728.1 | 3 - 4 | 264 | 112 | ||
NM_001383902.1 | 4 - 5 | 364 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |