Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267229.4 | NP_001254158.1 | 11 - 12 | 1964 | 108 | |
NM_001267230.4 | NP_001254159.1 | 10 - 11 | 1873 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |