Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267229.4 | NP_001254158.1 | 12 - 13 | 2064 | 88 | |
NM_001267230.4 | NP_001254159.1 | 11 - 12 | 1973 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |