Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383908.1 | 6 - 7 | 799 | 82 | ||
GenBank mRNA | AJ745071.1 | - | 6 - 7 | 784 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |