Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063539.6 | NP_495940.1 | 6 - 7 | 1798 | 78 | |
GenBank mRNA | AB035090.2 | - | 6 - 7 | 1796 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |