Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: let-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136358.4 | NP_001129830.1 | 4 - 5 | 796 | 93 | |
NM_001136359.5 | NP_001129831.1 | 6 - 7 | 994 | 93 | ||
NM_001313678.3 | NP_001300607.1 | 7 - 8 | 1021 | 93 | ||
NM_063561.5 | NP_495962.2 | 6 - 7 | 1068 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |