Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381562.1 | NP_001367038.1 | 8 - 9 | 1993 | 65 | |
NM_001383917.1 | 8 - 9 | 1984 | 65 | |||
NM_063593.6 | NP_495994.1 | 8 - 9 | 1985 | 65 | ||
GenBank mRNA | U86640.1 | - | 8 - 9 | 1976 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |