Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063693.5 | NP_496094.1 | 5 - 6 | 1048 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |