Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063700.4 | NP_496101.1 | 2 - 3 | 550 | 75 | |
NM_063701.4 | NP_496102.1 | 3 - 4 | 736 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |