Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063737.2 | NP_496138.1 | 5 - 6 | 717 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |