Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pro-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063870.6 | NP_496271.1 | 2 - 3 | 197 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |