Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064059.2 | NP_496460.2 | 2 - 3 | 321 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |