Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026907.4 | NP_001022078.1 | 2 - 3 | 111 | 82 | |
NM_001026908.6 | NP_001022079.1 | 2 - 3 | 117 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |