Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381616.2 | NP_001366714.1 | 6 - 7 | 631 | 62 | |
NM_001383961.1 | 7 - 8 | 882 | 62 | |||
GenBank mRNA | AF291846.1 | - | 6 - 7 | 628 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |