Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064178.6 | NP_496579.3 | 2 - 3 | 683 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |