Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sinh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064195.5 | NP_496596.3 | 3 - 4 | 972 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |