Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sop-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267411.2 | NP_001254340.1 | 5 - 6 | 1873 | 71 | |
NM_001267412.2 | NP_001254341.1 | 5 - 6 | 1867 | 71 | ||
NM_001267413.3 | NP_001254342.1 | 2 - 3 | 1187 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |