Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064226.5 | NP_496627.2 | 1 - 2 | 467 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |