Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064235.2 | NP_496636.1 | 2 - 3 | 718 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |