Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064258.2 | NP_496659.2 | 4 - 5 | 765 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |