Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcmo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064328.5 | NP_496729.2 | 5 - 6 | 1464 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |