Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027267.5 | NP_001022438.1 | 4 - 5 | 692 | 59 | |
NM_001373808.1 | NP_001359643.1 | 4 - 5 | 687 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |