Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064331.6 | NP_496732.1 | 3 - 4 | 1072 | 115 | |
GenBank mRNA | U15304.1 | - | 3 - 4 | 1071 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |