Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381654.1 | NP_001367183.1 | 6 - 7 | 1203 | 105 | |
NM_001381655.1 | NP_001366936.1 | 6 - 7 | 1216 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |