Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027102.6 | NP_001022273.1 | 9 - 10 | 1617 | 90 | |
NM_001381654.1 | NP_001367183.1 | 8 - 9 | 1494 | 90 | ||
NM_001381655.1 | NP_001366936.1 | 8 - 9 | 1507 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |