Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kbp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064592.6 | NP_496993.1 | 2 - 3 | 303 | 112 | |
GenBank mRNA | AF303250.1 | - | 2 - 3 | 295 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |