Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267504.3 | NP_001254433.1 | 2 - 3 | 213 | 88 | |
NM_001267505.2 | NP_001254434.1 | 2 - 3 | 211 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |