Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atgp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267506.3 | NP_001254435.1 | 8 - 9 | 1749 | 89 | |
NM_001267507.3 | NP_001254436.1 | 7 - 8 | 1608 | 89 | ||
NM_001267508.3 | NP_001254437.1 | 1 - 2 | 372 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |