Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027317.8 | NP_001022488.1 | 17 - 18 | 6225 | 56 | |
NM_001267510.3 | NP_001254439.1 | 18 - 19 | 5636 | 56 | ||
NM_001267511.3 | NP_001254440.1 | 16 - 17 | 5855 | 56 | ||
NM_001267513.3 | NP_001254442.1 | 16 - 17 | 5646 | 56 | ||
NM_001267514.4 | NP_001254443.1 | 17 - 18 | 5834 | 56 | ||
NM_001267515.3 | NP_001254444.1 | 18 - 19 | 6116 | 56 | ||
NM_001267516.3 | NP_001254445.1 | 18 - 19 | 6131 | 56 | ||
NM_001267517.3 | NP_001254446.1 | 19 - 20 | 6413 | 56 | ||
NM_001350388.3 | NP_001337294.1 | 29 | 8780 | 56 | ||
NM_001377905.1 | NP_001364664.1 | 28 - 29 | 8673 | 56 | ||
NM_064643.10 | NP_497044.3 | 19 - 20 | 6413 | 56 | ||
NM_064644.6 | NP_497045.1 | 17 - 18 | 5942 | 56 | ||
NM_064645.6 | NP_497046.1 | 19 - 20 | 6504 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |