Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ver-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064761.2 | NP_497162.2 | 6 - 7 | 2300 | 148 | |
GenBank mRNA | AF188751.1 | - | 6 - 7 | 2300 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |