Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083185.1 | NP_001076654.1 | 2 - 3 | 726 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |