Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368615.2 | NP_001355395.1 | 1 - 2 | 105 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |