Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iffb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065135.8 | NP_497536.2 | 8 - 9 | 2995 | 106 | |
GenBank mRNA | DQ867020.1 | - | 8 - 9 | 3017 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |