Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iffb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065135.8 | NP_497536.2 | 9 - 10 | 3153 | 98 | |
GenBank mRNA | DQ867020.1 | - | 9 - 10 | 3175 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |