Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: feh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027735.5 | NP_001022906.1 | 5 - 6 | 735 | 61 | |
NM_001373858.2 | NP_001360128.1 | 3 - 4 | 576 | 61 | ||
NM_001384041.1 | 4 - 5 | 696 | 61 | |||
NM_065177.6 | NP_497578.4 | 5 - 6 | 740 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |