Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129270.3 | NP_001122742.2 | 4 - 5 | 385 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |