Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267939.3 | NP_001254868.1 | 7 - 8 | 699 | 115 | |
NM_065293.6 | NP_497694.3 | 8 - 9 | 930 | 115 | ||
NM_065294.6 | NP_497695.3 | 8 - 9 | 931 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |