Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381809.2 | NP_001366800.1 | 2 - 3 | 428 | 71 | |
NM_001381810.1 | NP_001367205.1 | 2 - 3 | 418 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |