Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tir-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026080.5 | NP_001021251.2 | 5 - 6 | 1333 | 116 | |
NM_001312937.3 | NP_001299866.1 | 4 - 5 | 1211 | 116 | ||
NM_001381811.1 | NP_001368535.1 | 5 - 6 | 1049 | 116 | ||
NM_001381812.1 | NP_001367074.1 | 5 - 6 | 1333 | 116 | ||
NM_001384059.1 | 7 - 8 | 1358 | 116 | |||
NM_065383.4 | NP_497784.1 | 7 - 8 | 1375 | 116 | ||
GenBank mRNA | AY834227.1 | - | 5 - 6 | 1213 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |