Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001384060.1 | 3 - 4 | 560 | 75 | ||
GenBank mRNA | AB036738.1 | - | 3 - 4 | 560 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |