Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351863.2 | NP_001338832.1 | 4 - 5 | 677 | 111 | |
NM_065503.4 | NP_497904.1 | 4 - 5 | 650 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |