Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndg-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027447.6 | NP_001022618.1 | 4 - 5 | 591 | 101 | |
NM_001027448.4 | NP_001022619.1 | 4 - 5 | 591 | 101 | ||
GenBank mRNA | AF173373.1 | - | 4 - 5 | 589 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |