Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065517.5 | NP_497918.1 | 5 - 6 | 848 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |